Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSP93-III
Entrez Gene ID
824048
Gene Name
Clp ATPase
Gene Aliases
AT3G48870, ATCLPC, ATHSP93-III, ClpC2
Chromosome Location
Chr.3: 18122118 - 18126742 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203106.2 NP_001190035.1 9 - 10 1825 86
NM_001339385.1 NP_001325472.1 9 - 10 1967 86
NM_001339386.1 NP_001325473.1 10 - 11 1917 86
NM_114746.5 NP_566912.2 9 - 10 1867 86

View Full Product Details