Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
iqd21
Entrez Gene ID
824087
Gene Name
IQ-domain 21
Gene Aliases
AT3G49260, IQ-domain 21
Chromosome Location
Chr.3: 18262290 - 18266021 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203110.1 NP_001190039.1 3 - 4 604 72
NM_001339406.1 NP_001319716.1 3 - 4 489 72
NM_114785.4 NP_566917.1 3 - 4 713 72
NM_202676.1 NP_974405.1 2 - 3 647 72
GenBank mRNA AF361635.1 - 3 - 4 511 72
AY087861.1 - 3 - 4 552 72
AY484700.1 - 3 - 4 530 72
BT000602.1 - 2 - 3 403 72

View Full Product Details