Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CKA2
- Entrez Gene ID
-
824162
- Gene Name
- casein kinase II, alpha chain 2
- Gene Aliases
-
''casein kinase II, ALPHA CHAIN 2, AT3G50000, ATCKA2, ATPK15D, CASEIN KINASE II, alpha chain 2, alpha chain 2'', casein kinase II
- Chromosome Location
-
Chr.3: 18534450 - 18537073 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_114860.4 |
NP_190569.2 |
8 - 9 |
1122 |
62 |
GenBank mRNA |
BT000888.1 |
- |
8 - 9 |
1085 |
62 |
View Full Product Details