Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYCD3;3
Entrez Gene ID
824169
Gene Name
CYCLIN D3;3
Gene Aliases
AT3G50070, CYCLIN D3;3
Chromosome Location
Chr.3: 18565134 - 18566834 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114867.3 NP_190576.1 3 - 4 971 82
GenBank mRNA AY052665.1 - 3 - 4 882 82
AY063729.1 - 3 - 4 806 82
AY087498.1 - 3 - 4 917 82

View Full Product Details