Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CEN2
Entrez Gene ID
824198
Gene Name
centrin2
Gene Aliases
AT3G50360, ATCEN2, CEN1, CENTRIN 1, centrin2
Chromosome Location
Chr.3: 18674137 - 18675761 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114896.5 NP_190605.1 2 - 3 446 76
GenBank mRNA AJ009672.1 - 2 - 3 228 76
AK175359.1 - 2 - 3 217 76
AK175375.1 - 2 - 3 185 76
AK176828.1 - 2 - 3 185 76
AK176882.1 - 2 - 3 236 76
BT025522.1 - 1 - 2 162 76

View Full Product Details