Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRP2
Entrez Gene ID
824226
Gene Name
KIP-related protein 2
Gene Aliases
AT3G50630, ICK2, KIP-RELATED PROTEIN 2, KIP-related protein 2
Chromosome Location
Chr.3: 18800306 - 18801595 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339478.1 NP_001326590.1 1 - 2 526 80
NM_114923.4 NP_190632.1 1 - 2 526 80
GenBank mRNA AJ251851.1 - 1 - 2 324 80
AK176528.1 - 1 - 2 444 80
AK176728.1 - 1 - 2 444 80
AY088290.1 - 1 - 2 431 80
BT028944.1 - 1 - 2 324 80

View Full Product Details