Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS35C
Entrez Gene ID
824294
Gene Name
VPS35 homolog C
Gene Aliases
AT3G51310, VPS35 homolog C
Chromosome Location
Chr.3: 19044331 - 19049454 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114990.5 NP_190699.3 8 - 9 985 99
GenBank mRNA AB368490.1 - 8 - 9 852 99

View Full Product Details