Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RABG3A
Entrez Gene ID
826558
Gene Name
RAB GTPase homolog G3A
Gene Aliases
AT4G09720, ATRABG3A, F17A8.70, F17A8_70, RAB GTPase homolog G3A
Chromosome Location
Chr.4: 6132837 - 6135701 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340630.1 NP_001319888.1 3 - 4 675 158
NM_001340631.1 NP_001328831.1 3 - 4 687 158
NM_117040.4 NP_192710.1 4 - 5 566 158
NM_179016.1 NP_849347.1 2 - 3 334 158
GenBank mRNA AB071851.1 - 4 - 5 327 158
AK229171.1 - 4 - 5 457 158
BT029504.1 - 4 - 5 327 158
BT030039.1 - 2 - 3 225 158

View Full Product Details