Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HSD5
- Entrez Gene ID
-
826593
- Gene Name
- hydroxysteroid dehydrogenase 5
- Gene Aliases
-
AT4G10020, AtHSD5, T5L19.150, T5L19_150, hydroxysteroid dehydrogenase 5
- Chromosome Location
-
Chr.4: 6268273 - 6270434 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_117070.5 |
NP_192740.1 |
1 - 2 |
281 |
79 |
View Full Product Details