Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MTA
- Entrez Gene ID
-
826670
- Gene Name
- mRNAadenosine methylase
- Gene Aliases
-
AT4G10760, EMB1706, EMBRYO DEFECTIVE 1706, T12H20.6, T12H20_6, mRNAadenosine methylase
- Chromosome Location
-
Chr.4: 6619817 - 6623381 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_117144.4 |
NP_192814.1 |
3 - 4 |
1324 |
81 |
View Full Product Details