Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPA2
Entrez Gene ID
826712
Gene Name
SPA1-related 2
Gene Aliases
AT4G11110, SPA1-related 2, T22B4.90, T22B4_90
Chromosome Location
Chr.4: 6771605 - 6777225 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117181.4 NP_192849.4 2 - 3 1776 85

View Full Product Details