Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SR
Entrez Gene ID
826769
Gene Name
serine racemase
Gene Aliases
AT4G11640, ATSR, T5C23.70, T5C23_70, serine racemase
Chromosome Location
Chr.4: 7021422 - 7023318 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117233.3 NP_192901.2 4 - 5 1094 72
GenBank mRNA AB206823.1 - 4 - 5 746 72

View Full Product Details