Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HAP2
Entrez Gene ID
826777
Gene Name
hapless 2
Gene Aliases
AT4G11720, GCS1, GENERATIVE CELL-SPECIFIC 1, HAPLESS 2, T5C23.150, T5C23_150
Chromosome Location
Chr.4: 7063255 - 7066878 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117241.2 NP_192909.2 10 - 11 1210 108

View Full Product Details