Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAY1
Entrez Gene ID
826779
Gene Name
Ubiquitin-like superfamily protein
Gene Aliases
AT4G11740, T5C23.170, T5C23_170
Chromosome Location
Chr.4: 7071672 - 7075537 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340732.1 NP_001328454.1 4 - 5 923 95
NM_117243.4 NP_567380.2 4 - 5 1018 95

View Full Product Details