Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDX14
Entrez Gene ID
826805
Gene Name
nudix hydrolase homolog 14
Gene Aliases
ARABIDOPSIS THALIANA NUDIX HYDROLASE HOMOLOG 14, AT4G11980, ATNUDT14, ATNUDX14, F16J13.50, F16J13_50, nudix hydrolase homolog 14
Chromosome Location
Chr.4: 7183650 - 7185581 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117268.3 NP_567384.1 2 - 3 348 118
GenBank mRNA AJ748742.1 - 2 - 3 332 118
AY052271.1 - 2 - 3 341 118
BT001146.1 - 2 - 3 318 118

View Full Product Details