Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CMT2
Entrez Gene ID
827640
Gene Name
chromomethylase 2
Gene Aliases
AT4G19020, F13C5.190, F13C5_190, chromomethylase 2
Chromosome Location
Chr.4: 10414428 - 10421265 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341318.1 NP_001329724.1 16 - 17 3027 79
NM_118020.5 NP_193637.2 15 - 16 3206 79

View Full Product Details