Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCE2
Entrez Gene ID
827661
Gene Name
RNAse l inhibitor protein 2
Gene Aliases
ARABIDOPSIS THALIANA RNASE L INHIBITOR PROTEIN 2, AT4G19210, ATP-binding cassette E2, ATRLI2, RLI2, RNAse l inhibitor protein 2, T18B16.180, T18B16_180
Chromosome Location
Chr.4: 10501123 - 10505217 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118041.5 NP_193656.2 9 - 10 1391 69
GenBank mRNA AK226939.1 - 9 - 10 1298 69
AY099697.1 - 9 - 10 1311 69
BT000298.1 - 8 - 9 1187 69

View Full Product Details