Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEL1
Entrez Gene ID
828876
Gene Name
Eukaryotic release factor 1 (eRF1) family protein
Gene Aliases
AT4G27650, PELOTA, T29A15.140, T29A15_140
Chromosome Location
Chr.4: 13803268 - 13808031 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341876.1 NP_001329759.1 8 - 9 780 95
NM_001341877.1 NP_001329760.1 7 - 8 938 95
NM_118901.5 NP_194495.3 8 - 9 769 95
GenBank mRNA AF069497.1 - 8 - 9 664 95
AY074298.1 - 8 - 9 643 95
AY096569.1 - 7 - 8 463 95

View Full Product Details