Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sks10
Entrez Gene ID
828925
Gene Name
SKU5 similar 10
Gene Aliases
AT4G28090, SKU5 similar 10, SKU5 similar 10, T13J8.200, T13J8_200
Chromosome Location
Chr.4: 13961888 - 13964229 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118949.1 NP_194538.1 5 - 6 1349 63
GenBank mRNA DQ446876.1 - 5 - 6 1349 63

View Full Product Details