Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EXPB3
Entrez Gene ID
828940
Gene Name
expansin B3
Gene Aliases
AT4G28250, ATEXPB3, ATHEXP BETA 1.6, F26K10.130, F26K10_130, expansin B3
Chromosome Location
Chr.4: 13999625 - 14002124 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084985.1 NP_001078454.1 1 - 2 282 80
NM_001341907.1 NP_001329906.1 1 - 2 282 80
NM_001341908.1 NP_001329907.1 1 - 2 282 80
NM_118965.4 NP_567803.1 1 - 2 337 80
GenBank mRNA AY039986.1 - 1 - 2 254 80
AY133801.1 - 1 - 2 158 80

View Full Product Details