Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDX19
Entrez Gene ID
832129
Gene Name
nudix hydrolase homolog 19
Gene Aliases
ARABIDOPSIS THALIANA NUDIX HYDROLASE HOMOLOG 19, AT5G20070, ATNUDT19, ATNUDX19, F28I16.220, F28I16_220, nudix hydrolase homolog 19
Chromosome Location
Chr.5: 6779841 - 6782501 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122014.4 NP_197507.1 2 - 3 787 81
GenBank mRNA AK226922.1 - 2 - 3 751 81
AY049280.1 - 2 - 3 768 81
AY090287.1 - 2 - 3 735 81

View Full Product Details