Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPX1
Entrez Gene ID
832137
Gene Name
SPX domain-containing protein 1
Gene Aliases
ARABIDOPSIS THALIANA SPX DOMAIN GENE 1, ARABIDOPSIS THALIANA SPX DOMAIN GENE 1, AT5G20150, ATSPX1, F5O24.40, F5O24_40, SPX domain gene 1, SPX domain gene 1
Chromosome Location
Chr.5: 6802282 - 6803655 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122022.3 NP_197515.1 1 - 2 411 153
GenBank mRNA AY075605.1 - 1 - 2 306 153
BT000868.1 - 1 - 2 264 153

View Full Product Details