Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DD46
Entrez Gene ID
838806
Gene Name
Galactosyltransferase family protein
Gene Aliases
AT1G22015
Chromosome Location
Chr.1: 7750994 - 7753519 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102051.3 NP_564154.1 8 - 9 901 106
GenBank mRNA AK226416.1 - 8 - 9 902 106
KJ138995.1 - 8 - 9 807 106

View Full Product Details