Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DCL4
Entrez Gene ID
832154
Gene Name
dicer-like 4
Gene Aliases
AT5G20320, ATDCL4, DICER-LIKE 4, F5O24.210, F5O24_210, dicer-like 4
Chromosome Location
Chr.5: 6859212 - 6869307 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203419.2 NP_001190348.1 2 - 3 799 57
NM_122039.5 NP_197532.3 2 - 3 758 57
GenBank mRNA DQ118423.1 - 2 - 3 758 57

View Full Product Details