Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AL5
Entrez Gene ID
832173
Gene Name
alfin-like 5
Gene Aliases
AT5G20510, F7C8.100, F7C8_100, alfin-like 5
Chromosome Location
Chr.5: 6939813 - 6943011 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343672.1 NP_001331722.1 3 - 4 585 133
NM_001343673.1 NP_001331723.1 3 - 4 426 133
NM_122058.4 NP_197551.2 3 - 4 529 133
GenBank mRNA BT015937.1 - 3 - 4 407 133
BT020505.1 - 3 - 4 364 133

View Full Product Details