Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WAV2
Entrez Gene ID
832174
Gene Name
alpha/beta-Hydrolases superfamily protein
Gene Aliases
AT5G20520, F7C8.110, F7C8_110, WAVY GROWTH 2
Chromosome Location
Chr.5: 6942955 - 6946514 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122059.4 NP_568395.1 7 - 8 1115 69
GenBank mRNA AB182157.1 - 7 - 8 1035 69
AY080751.1 - 7 - 8 1050 69
AY087569.1 - 7 - 8 1046 69
AY114009.1 - 7 - 8 916 69

View Full Product Details