Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MS3
Entrez Gene ID
832223
Gene Name
methionine synthase 3
Gene Aliases
AT5G20980, ATMS3, F22D1.150, F22D1_150, methionine synthase 3
Chromosome Location
Chr.5: 7122800 - 7128412 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085142.1 NP_001078611.1 7 - 8 1353 65
NM_122107.4 NP_197598.2 7 - 8 1406 65
GenBank mRNA AK229291.1 - 7 - 8 1353 65

View Full Product Details