Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
QQT1
Entrez Gene ID
832298
Gene Name
P-loop containing nucleoside triphosphate hydrolases superfamily protein
Gene Aliases
AT5G22370, EMB1705, EMBRYO DEFECTIVE 1705, MWD9.17, MWD9_17, QUATRE-QUART 1
Chromosome Location
Chr.5: 7406030 - 7408754 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125789.1 NP_001119261.1 7 - 8 655 125
NM_001343725.1 NP_001332350.1 6 - 7 918 125
NM_122142.4 NP_197629.2 7 - 8 725 125
GenBank mRNA AK221541.1 - 7 - 8 624 125
AX765846.1 - 6 - 7 553 125

View Full Product Details