Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MGT10
- Entrez Gene ID
-
832346
- Gene Name
- magnesium (Mg) transporter 10
- Gene Aliases
-
AT5G22830, ATMGT10, GMN10, MAGNESIUM TRANSPORTER 10, MRN17.6, MRN17_6, MRS2-11, magnesium (Mg) transporter 10
- Chromosome Location
-
Chr.5: 7627597 - 7630993 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_122188.3 |
NP_568424.1 |
13 - 14 |
1490 |
141 |
View Full Product Details