Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HTB2
Entrez Gene ID
832352
Gene Name
histone B2
Gene Aliases
AT5G22880, H2B, HISTONE H2B, MRN17.11, MRN17_11, histone B2
Chromosome Location
Chr.5: 7651868 - 7652714 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122194.3 NP_197679.1 1 - 1 654 156

View Full Product Details