The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_122236.3 | NP_568428.1 | 7 - 8 | 1337 | 75 |
GenBank mRNA | AF454729.1 | - | 7 - 8 | 1176 | 75 |
AK228165.1 | - | 7 - 8 | 1337 | 75 | |
BT026524.1 | - | 7 - 8 | 1176 | 75 |