The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_122249.4 | NP_568431.1 | 5 - 6 | 681 | 73 |
GenBank mRNA | AK118688.1 | - | 5 - 6 | 499 | 73 |