Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PDR2
- Entrez Gene ID
-
832428
- Gene Name
- phosphate deficiency response 2
- Gene Aliases
-
AT5G23630, MALE GAMETOGENESIS IMPAIRED ANTHERS, MIA, MQM1.11, MQM1_11, PI DEFICIENCY RESPONSE 2, phosphate deficiency response 2
- Chromosome Location
-
Chr.5: 7960478 - 7968197 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_122268.5 |
NP_197752.1 |
8 - 9 |
1474 |
73 |
View Full Product Details