Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MIND
Entrez Gene ID
832467
Gene Name
septum site-determining protein (MIND)
Gene Aliases
ACCUMULATION AND REPLICATION OF CHLOROPLASTS 11, ARC11, AT5G24020, ATMIND1, MZF18.10, MZF18_10
Chromosome Location
Chr.5: 8116665 - 8118087 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122307.3 NP_197790.1 1 - 1 1249 75
GenBank mRNA BT002027.1 - 1 - 1 1234 75

View Full Product Details