Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YSL2
Entrez Gene ID
832509
Gene Name
YELLOW STRIPE like 2
Gene Aliases
AT5G24380, ATYSL2, K16H17.9, K16H17_9, YELLOW STRIPE LIKE 2, YELLOW STRIPE like 2
Chromosome Location
Chr.5: 8323895 - 8326705 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122346.3 NP_197826.2 4 - 5 1138 102
GenBank mRNA AY515561.1 - 4 - 5 1090 102
AY648977.1 - 4 - 5 935 102

View Full Product Details