Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FPF1
Entrez Gene ID
832555
Gene Name
flowering promoting factor 1
Gene Aliases
ARABIDOPSIS FLOWERING PROMOTING FACTOR 1, AT5G24860, ATFPF1, F6A4.70, F6A4_70, FLOWERING PROMOTING FACTOR 1
Chromosome Location
Chr.5: 8541558 - 8542473 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343900.1 NP_001330701.1 1 - 1 760 131
NM_001343901.1 NP_001330702.1 1 - 1 605 131
NM_122395.5 NP_197868.1 1 - 1 567 131
GenBank mRNA Y11988.1 - - 506 131

View Full Product Details