Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RD22
Entrez Gene ID
832636
Gene Name
BURP domain-containing protein
Gene Aliases
AT5G25610, ATRD22, RESPONSIVE TO DESSICATION 22, T14C9.150, T14C9_150
Chromosome Location
Chr.5: 8914122 - 8916856 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122472.4 NP_197943.1 2 - 3 314 75
GenBank mRNA AY060560.1 - 2 - 3 180 75
AY090244.1 - 2 - 3 180 75
BT009697.1 - 2 - 3 142 75

View Full Product Details