Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YUC6
Entrez Gene ID
832638
Gene Name
Flavin-binding monooxygenase family protein
Gene Aliases
AT5G25620, AtYUC6, T14C9.160, T14C9_160, YUCCA6
Chromosome Location
Chr.5: 8934941 - 8938768 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203470.1 NP_001190399.2 2 - 3 1477 132
NM_122473.3 NP_197944.2 2 - 3 1487 132
GenBank mRNA AY065229.1 - 2 - 3 1385 132
AY113851.1 - 2 - 3 943 132

View Full Product Details