Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RLP52
Entrez Gene ID
832660
Gene Name
receptor like protein 52
Gene Aliases
AT5G25910, AtRLP52, T1N24.21, T1N24_21, receptor like protein 52
Chromosome Location
Chr.5: 9038860 - 9041377 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122492.1 NP_197963.1 1 - 2 1274 108

View Full Product Details