Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY50
Entrez Gene ID
832686
Gene Name
WRKY DNA-binding protein 50
Gene Aliases
ARABIDOPSIS THALIANA WRKY DNA-BINDING PROTEIN 50, AT5G26170, ATWRKY50, T19G15.20, T19G15_20, WRKY DNA-binding protein 50
Chromosome Location
Chr.5: 9146879 - 9148391 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122518.3 NP_197989.2 1 - 2 522 116
GenBank mRNA AB493759.1 - 1 - 2 259 116
AY071847.1 - 1 - 2 259 116

View Full Product Details