Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IREG3
Entrez Gene ID
832740
Gene Name
iron-regulated protein 3
Gene Aliases
AT5G26820, ATIREG3, F2P16.6, F2P16_6, IRON REGULATED 3, MAR1, MULTIPLE ANTIBIOTIC RESISTANCE 1, RTS3, iron-regulated protein 3
Chromosome Location
Chr.5: 9433569 - 9437158 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122564.5 NP_198034.2 1 - 2 461 122
GenBank mRNA AY062606.1 - 1 - 2 413 122
AY128787.1 - 1 - 2 172 122

View Full Product Details