Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SULTR1;3
Entrez Gene ID
838820
Gene Name
sulfate transporter 1;3
Gene Aliases
AT1G22150, F2E2.22, F2E2_22, SULFATE TANSPORTER SULTR1;3, sulfate transporter 1;3
Chromosome Location
Chr.1: 7817609 - 7821576 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332538.1 NP_001319061.1 7 - 8 1141 75
NM_001332539.1 NP_001321054.1 7 - 8 1015 75
NM_001332540.1 NP_001321055.1 7 - 8 1015 75
NM_102065.2 NP_564159.1 6 - 7 940 75
GenBank mRNA AB049624.1 - 6 - 7 940 75

View Full Product Details