Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZYP1b
Entrez Gene ID
838834
Gene Name
Myosin heavy chain-related protein
Gene Aliases
AT1G22275, ZYP1
Chromosome Location
Chr.1: 7867067 - 7872715 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332545.1 NP_001323354.1 13 - 14 2312 66
NM_102078.3 NP_564164.1 13 - 14 2346 66
GenBank mRNA DQ028765.1 - 13 - 14 2280 66

View Full Product Details