Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FRO3
Entrez Gene ID
838910
Gene Name
ferric reduction oxidase 3
Gene Aliases
AT1G23020, ATFRO3, F19G10.4, F19G10_4, FERRIC REDUCTION OXIDASE 3, ferric reduction oxidase 3
Chromosome Location
Chr.1: 8149868 - 8153796 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332575.1 NP_001319065.1 4 - 5 501 123
NM_001332576.1 NP_001322197.1 3 - 4 605 123
NM_001332577.1 NP_001322198.1 2 - 3 611 123
NM_102150.4 NP_173715.2 3 - 4 507 123

View Full Product Details