Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TAR1
- Entrez Gene ID
-
838941
- Gene Name
- tryptophan aminotransferase related 1
- Gene Aliases
-
AT1G23320, F26F24.17, F26F24_17, tryptophan aminotransferase related 1
- Chromosome Location
-
Chr.1: 8273423 - 8275350 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_102181.1 |
NP_173746.1 |
1 - 2 |
89 |
109 |
View Full Product Details