Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LGT9
Entrez Gene ID
839030
Gene Name
Nucleotide-diphospho-sugar transferases superfamily protein
Gene Aliases
AT1G24170, F3I6.10, F3I6_10, GALACTURONOSYLTRANSFERASE-LIKE 8, GATL8
Chromosome Location
Chr.1: 8557215 - 8559066 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102263.3 NP_173827.1 1 - 1 1684 171
GenBank mRNA AK119069.1 - 1 - 1 1450 171

View Full Product Details