Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- KCS5
- Entrez Gene ID
-
839131
- Gene Name
- 3-ketoacyl-CoA synthase 5
- Gene Aliases
-
3-ketoacyl-CoA synthase 5, AT1G25450, CER60, ECERIFERUM 60, F2J7.9, F2J7_9
- Chromosome Location
-
Chr.1: 8938387 - 8940613 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_102356.4 |
NP_173916.1 |
1 - 2 |
1362 |
86 |
View Full Product Details