Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PFT1
Entrez Gene ID
839141
Gene Name
phytochrome and flowering time regulatory protein (PFT1)
Gene Aliases
AT1G25540, F2J7.4, F2J7_4, MED25, PHYTOCHROME AND FLOWERING TIME 1, mediator 25
Chromosome Location
Chr.1: 8969065 - 8974815 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084127.1 NP_001077596.1 5 - 6 787 120
NM_102365.5 NP_173925.3 6 - 7 973 120
GenBank mRNA AY170377.1 - 6 - 7 604 120

View Full Product Details