Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PERK10
- Entrez Gene ID
-
839156
- Gene Name
- proline-rich extensin-like receptor kinase 10
- Gene Aliases
-
AT1G26150, ATPERK10, AtPERK10, F28B23.17, F28B23_17, proline-rich extensin-like receptor kinase 10
- Chromosome Location
-
Chr.1: 9039628 - 9043855 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_102380.4 |
NP_173940.2 |
7 - 8 |
3070 |
79 |
View Full Product Details