Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRF12
Entrez Gene ID
839189
Gene Name
general regulatory factor 12
Gene Aliases
AT1G26480, GF14 IOTA, T1K7.15, T1K7_15, general regulatory factor 12
Chromosome Location
Chr.1: 9156279 - 9158093 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102411.3 NP_564249.1 1 - 2 330 69
GenBank mRNA AF335544.1 - 1 - 2 82 69
AK117907.1 - 1 - 2 186 69
BT006230.1 - 1 - 2 82 69

View Full Product Details